Assay Design



Gene Transcripts

Gene Symbol
Clic
Entrez Gene ID
32349
Gene Name
Chloride intracellular channel
Gene Aliases
CG10997, CLIC, DCLIC, DmCLIC, Dmel\CG10997, Dmel_CG10997, clic, l(1)G0053, l(1)G0472
Chromosome Location
Chr.X: 13812971 - 13822307 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0030529
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001272608.2 NP_001259537.2 1 - 2 389 86
NM_132700.4 NP_572928.1 1 - 2 389 86
GenBank mRNA AY058721.1 - 1 - 2 406 86

View Full Product Details