Assay Design
Gene Transcripts
- Gene Symbol
- mamo
- Entrez Gene ID
-
32353
- Gene Name
- maternal gene required for meiosis
- Gene Aliases
-
CG11071, CG11072, CG11075, CG11082, CG32606, CG32611, CG34346, CG44132, CG45477, CR44656, Dmel\CG45477, Dmel_CG45477, lincRNA.990
- Chromosome Location
-
Chr.X: 13846116 - 13990527 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0267033
View Full Product Details