Assay Design



Gene Transcripts

Gene Symbol
tay
Entrez Gene ID
32547
Gene Name
tay bridge
Gene Aliases
CG9056, Dmel\CG9056, Dmel_CG9056, XI, dAUTS2, l(1)13Ff, l(1)14Ac, l(1)XI
Chromosome Location
Chr.X: 15876368 - 15887311 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0260938
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001272656.2 NP_001259585.1 6 - 7 2951 133
NM_001272657.2 NP_001259586.1 6 - 7 2871 133
NM_001298376.1 NP_001285305.1 6 - 7 2973 133
NM_132858.3 NP_573086.2 6 - 7 2976 133
GenBank mRNA AY118524.1 - 6 - 7 2951 133

View Full Product Details