Assay Design



Gene Transcripts

Gene Symbol
Teh2
Entrez Gene ID
38502
Gene Name
tipE homolog 2
Gene Aliases
CG15004, Dmel\CG15004, Dmel_CG15004, TEH2
Chromosome Location
Chr.3L: 4178980 - 4188023 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0035505
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169874.3 NP_001163345.1 3 - 4 1900 62
NM_139610.4 NP_647867.1 2 - 3 512 62
GenBank mRNA BT050528.1 - 3 - 4 1901 62
DQ139958.1 - 2 - 3 477 62

View Full Product Details