Assay Design



Gene Transcripts

Gene Symbol
DopEcR
Entrez Gene ID
38539
Gene Name
Dopamine/Ecdysteroid receptor
Gene Aliases
CG18314, DmDopEcR, Dmel\CG18314, Dmel_CG18314, DopECR, anon-WO0170980.139, anon-WO0170980.140, dop
Chromosome Location
Chr.3L: 4367649 - 4380387 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0035538
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014559.3 NP_001014559.1 3 - 4 1744 76
NM_001014560.2 NP_001014560.1 4 - 5 2198 76
NM_139640.3 NP_647897.2 4 - 5 1820 76

View Full Product Details