Assay Design



Gene Transcripts

Gene Symbol
DIP-delta
Entrez Gene ID
5740816
Gene Name
Dpr-interacting protein delta
Gene Aliases
34391, CG15214, CG34391, CG4814, CT15465, CT35147, Dmel\CG34391, Dmel_CG34391
Chromosome Location
Chr.3L: 5424109 - 5442399 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0085420
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104038.4 NP_001097508.2 10 - 11 1588 75
NM_001144414.3 NP_001137886.1 3 - 4 331 75
NM_001274512.2 NP_001261441.1 10 - 11 1588 75
GenBank mRNA BT023547.1 - 3 - 4 291 75

View Full Product Details