Assay Design



Gene Transcripts

Gene Symbol
lin-28
Entrez Gene ID
38639
Gene Name
CG17334-PA
Gene Aliases
BcDNA:RE05342, CG17334, Dmel\CG17334, Dmel_CG17334, EP(3)0915, LIN-28, Lin-28, Lin28, lin28
Chromosome Location
Chr.3L: 5654681 - 5657384 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0035626
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139726.2 NP_647983.1 2 - 3 503 57
GenBank mRNA AF521096.2 - 2 - 3 500 57
AY118360.1 - 2 - 3 500 57

View Full Product Details