Assay Design
Gene Transcripts
- Gene Symbol
- MED4
- Entrez Gene ID
-
38799
- Gene Name
- Mediator complex subunit 4
- Gene Aliases
-
CG8609, Dmel\CG8609, Dmel_CG8609, Med4, Med4/TRAP36, Trap36, dTRAP36, dp34, med4, p34
- Chromosome Location
-
Chr.3L: 7249529 - 7250527 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0035754
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_139837.3 |
NP_648094.1 |
1 - 2 |
317 |
57 |
GenBank mRNA |
AY061502.1 |
- |
1 - 2 |
292 |
57 |
View Full Product Details