Assay Design
Gene Transcripts
- Gene Symbol
- ppk26
- Entrez Gene ID
-
38835
- Gene Name
- pickpocket 26
- Gene Aliases
-
CG8546, Dmel\CG8546, Dmel_CG8546, bba
- Chromosome Location
-
Chr.3L: 7456149 - 7458608 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0035785
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_139868.3 |
NP_648125.3 |
2 - 3 |
846 |
83 |
View Full Product Details