Assay Design
Gene Transcripts
- Gene Symbol
- Srp9
- Entrez Gene ID
-
38883
- Gene Name
- Signal recognition particle 9
- Gene Aliases
-
CG8268, Dmel\CG8268, Dmel_CG8268
- Chromosome Location
-
Chr.3L: 7936373 - 7937258 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0035827
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_139906.3 |
NP_648163.1 |
1 - 2 |
248 |
70 |
GenBank mRNA |
AY071180.1 |
- |
1 - 2 |
246 |
70 |
View Full Product Details