Assay Design
Gene Transcripts
- Gene Symbol
- eIF4E3
- Entrez Gene ID
-
38923
- Gene Name
- eukaryotic translation initiation factor 4E3
- Gene Aliases
-
CG8023, Dmel\CG8023, Dmel_CG8023, anon-WO0140519.227, eIF4E, eIF4E-3, elF4E-3, l(3)0139, l(3)L0139
- Chromosome Location
-
Chr.3L: 8229224 - 8230568 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0265089
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_139937.2 |
NP_648194.1 |
1 - 2 |
211 |
73 |
GenBank mRNA |
AY122090.1 |
- |
1 - 2 |
151 |
73 |
View Full Product Details