Assay Design
Gene Transcripts
- Gene Symbol
- mtrm
- Entrez Gene ID
-
38958
- Gene Name
- matrimony
- Gene Aliases
-
CG18543, D52, Dmel\CG18543, Dmel_CG18543, Mtrm, anon-D52, anonD52
- Chromosome Location
-
Chr.3L: 8413012 - 8414103 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0010431
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_139966.3 |
NP_648223.1 |
1 - 1 |
682 |
95 |
GenBank mRNA |
AY061524.1 |
- |
1 - 1 |
637 |
95 |
View Full Product Details