Assay Design
Gene Transcripts
- Gene Symbol
- ZC3H3
- Entrez Gene ID
-
38968
- Gene Name
- CG6694-PA
- Gene Aliases
-
CG6694, Dmel\CG6694, Dmel_CG6694, dZC3H3
- Chromosome Location
-
Chr.3L: 8507469 - 8509763 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0035900
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_139973.3 |
NP_648230.1 |
2 - 3 |
1330 |
69 |
GenBank mRNA |
BT010061.1 |
- |
2 - 3 |
1315 |
69 |
View Full Product Details