Assay Design
Gene Transcripts
- Gene Symbol
- Gpdh1
- Entrez Gene ID
-
33824
- Gene Name
- Glycerol-3-phosphate dehydrogenase 1
- Gene Aliases
-
CG9042, DROGPDHA, DmG3PDH, Dmel\CG9042, Dmel_CG9042, G3PDH, G3pdh, GAPDH, GPD, GPDA, GPDH, GPDH-1, Gdh, Gpd, Gpdh, alpha-GPD, alpha-GPDH, alpha-GPDH-1, alpha-Gpdh, alphaGPDH, alphaGpd, alphaGpdh, alphaGpdh-1, dGpdh, gpdh, gpdh-1, sn-Gpdh
- Chromosome Location
-
Chr.2L: 5943643 - 5948968 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0001128
View Full Product Details