Assay Design
Gene Transcripts
- Gene Symbol
- Mal-A1
- Entrez Gene ID
-
35824
- Gene Name
- Maltase A1
- Gene Aliases
-
CG8696, Dmel\CG8696, Dmel_CG8696, Drome_A1, H, LvpH, V, lvpH, mal_A1
- Chromosome Location
-
Chr.2R: 8449664 - 8451904 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0002570
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_057279.5 |
NP_476627.3 |
1 - 2 |
1350 |
92 |
View Full Product Details