Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001273027.1 | NP_001259956.1 | 2 - 3 | 549 | 51 |
NM_001273028.1 | NP_001259957.1 | 2 - 3 | 556 | 51 | |
NM_001273029.1 | NP_001259958.1 | 2 - 3 | 554 | 51 | |
NM_001273030.1 | NP_001259959.1 | 2 - 3 | 582 | 51 | |
NM_057328.4 | NP_476676.1 | 1 - 2 | 175 | 51 | |
GenBank mRNA | BT023910.1 | - | - | 185 | 51 |