Assay Design
Gene Transcripts
- Gene Symbol
- Pig1
- Entrez Gene ID
-
31303
- Gene Name
- Pre-intermoult gene 1
- Gene Aliases
-
CG10790, Dmel\CG10790, Dmel_CG10790, EG:96G10.1, Pig-1, gsg, pig1
- Chromosome Location
-
Chr.X: 3248600 - 3249289 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0003086
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_057329.4 |
NP_476677.2 |
1 - 1 |
660 |
123 |
View Full Product Details