Assay Design



Gene Transcripts

Gene Symbol
Shaw
Entrez Gene ID
33599
Gene Name
Shaker cognate w
Gene Aliases
CG15419, CG2822, Dmel\CG2822, Dmel_CG2822, K[[v]]3, Kv3, Kv3.1, Shaw2, dShaw, dShaw2, shaw
Chromosome Location
Chr.2L: 3718185 - 3729145 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0003386
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144310.1 NP_001137782.1 5 - 6 1388 101
NM_001273071.1 NP_001260000.1 6 - 7 1646 101
NM_001273072.1 NP_001260001.1 8 - 9 1800 101
NM_057373.3 NP_476721.1 8 - 9 1800 101
NM_164554.3 NP_722937.2 8 - 9 1800 101
GenBank mRNA AY071475.1 - 8 - 9 1798 101

View Full Product Details