Assay Design



Gene Transcripts

Gene Symbol
sli
Entrez Gene ID
36746
Gene Name
slit
Gene Aliases
CG33464, CG43758, CG8355, CT21700, CT37068, Dmel\CG43758, Dmel_CG43758, Sli, Slit, dSlit, fus3, l(2)05248, wdh
Chromosome Location
Chr.2R: 15869495 - 15922118 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0264089
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103863.3 NP_001097333.1 5 - 6 1761 66
NM_001103864.3 NP_001097334.1 5 - 6 990 66
NM_001274088.1 NP_001261017.1 6 - 7 1833 66
NM_057379.4 NP_476727.1 6 - 7 1833 66
NM_057380.4 NP_476728.1 5 - 6 1761 66
NM_057381.4 NP_476729.1 6 - 7 1823 66

View Full Product Details