Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001103863.3 | NP_001097333.1 | 8 - 9 | 2281 | 72 |
NM_001103864.3 | NP_001097334.1 | 8 - 9 | 1510 | 72 | |
NM_001274088.1 | NP_001261017.1 | 9 - 10 | 2353 | 72 | |
NM_057379.4 | NP_476727.1 | 9 - 10 | 2353 | 72 | |
NM_057380.4 | NP_476728.1 | 8 - 9 | 2281 | 72 | |
NM_057381.4 | NP_476729.1 | 9 - 10 | 2343 | 72 |