Assay Design
Gene Transcripts
- Gene Symbol
- Klp61F
- Entrez Gene ID
-
38135
- Gene Name
- Kinesin-like protein at 61F
- Gene Aliases
-
6345, CG9191, CG9191-PA, DmKLP61F, DmKRP130, DmKlp61F, Dmel\CG9191, Dmel_CG9191, KIF 8, KIF11, KLP2, KLP61, KLP61F, KLP61f, KRP130, KRP[[(130)]], KRP[[130]], Kin-5, Klp 61F, Klp2, Klp61F/KRP130, Klp61f, Krp130, klp61F, klp61f, l(3)07012, l(3)6D283, l(3)Klp61F, urc, urcn
- Chromosome Location
-
Chr.3L: 1241568 - 1245910 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0004378
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_057470.5 |
NP_476818.1 |
4 - 5 |
3199 |
63 |
GenBank mRNA |
AY069442.1 |
- |
4 - 5 |
3194 |
63 |
View Full Product Details