Assay Design



Gene Transcripts

Gene Symbol
ph-p
Entrez Gene ID
31181
Gene Name
polyhomeotic proximal
Gene Aliases
CG18412, CG18414, Dmel\CG18412, Dmel_CG18412, EG:87B1.5, PH, PH-p, PHP, Ph, Ph-p, PhP, dPh, ph, ph-P, ph-proximal, ph[P], ph[p], phm, php
Chromosome Location
Chr.X: 2126210 - 2138371 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0004861
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001297873.1 NP_001284802.1 2 - 3 368 66
NM_057523.5 NP_476871.2 1 - 2 613 66

View Full Product Details