Assay Design



Gene Transcripts

Gene Symbol
piwi
Entrez Gene ID
34521
Gene Name
P-element induced wimpy testis
Gene Aliases
CG6122, Dmel\CG6122, Dmel_CG6122, PIWI, Piwi
Chromosome Location
Chr.2L: 10982205 - 10987420 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0004872
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001298896.1 NP_001285825.1 5 - 6 1440 74
NM_057527.4 NP_476875.1 5 - 6 1440 74

View Full Product Details