Assay Design
Gene Transcripts
- Gene Symbol
- Brd
- Entrez Gene ID
-
39620
- Gene Name
- Bearded
- Gene Aliases
-
Brd-C, CG3096, DmelCG3096, Dmel\CG3096, Dmel_CG3096, brd
- Chromosome Location
-
Chr.3L: 14972667 - 14973198 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0000216
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_057541.5 |
NP_476889.1 |
1 - 1 |
408 |
62 |
View Full Product Details