Assay Design



Gene Transcripts

Gene Symbol
ple
Entrez Gene ID
38746
Gene Name
pale
Gene Aliases
CG10118, DH65B, DTH, Dmel\CG10118, Dmel_CG10118, Pale, Ple, TH, Th, TyrH, VII, dTH1, dTH65B, pale/ple, th
Chromosome Location
Chr.3L: 6713356 - 6719525 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0005626
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300026.1 NP_001286955.1 2 - 3 629 117
NM_057550.4 NP_476898.1 2 - 3 629 117
GenBank mRNA BT044563.1 - 2 - 3 627 117

View Full Product Details