Assay Design



Gene Transcripts

Gene Symbol
pigeon
Entrez Gene ID
35200
Gene Name
CG10739-PA
Gene Aliases
CG10739, Dmel\CG10739, Dmel_CG10739, lio, pio
Chromosome Location
Chr.2L: 19186043 - 19189938 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0010309
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001201915.2 NP_001188844.1 2 - 3 556 66
NM_057598.5 NP_476946.1 2 - 3 556 66
GenBank mRNA AY061465.1 - 2 - 3 545 66

View Full Product Details