Assay Design



Gene Transcripts

Gene Symbol
CalpA
Entrez Gene ID
37232
Gene Name
Calpain-A
Gene Aliases
CG18152, CG7563, Dmel\CG7563, Dmel_CG7563, EST C, calpA
Chromosome Location
Chr.2R: 19425113 - 19430564 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0012051
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103908.2 NP_001097378.1 7 - 8 1049 70
NM_001299684.1 NP_001286613.1 8 - 9 1094 70
NM_057699.5 NP_477047.1 7 - 8 957 70
NM_057700.4 NP_477048.1 7 - 8 1049 70
GenBank mRNA AY051678.1 - 6 - 7 933 70

View Full Product Details