Assay Design
Gene Transcripts
- Gene Symbol
- Odc2
- Entrez Gene ID
-
35767
- Gene Name
- Ornithine decarboxylase 2
- Gene Aliases
-
CG8719, Dmel\CG8719, Dmel_CG8719, dodc2
- Chromosome Location
-
Chr.2R: 8007246 - 8009062 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0013308
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_057705.4 |
NP_477053.2 |
5 - 6 |
1060 |
92 |
View Full Product Details