Assay Design



Gene Transcripts

Gene Symbol
gcm
Entrez Gene ID
34277
Gene Name
glial cells missing
Gene Aliases
CG12245, Dmel\CG12245, Dmel_CG12245, GCM, GCM1, GCMa, Gcm, N7-4, glide, glide/gcm, l(2)N7-4, ucc
Chromosome Location
Chr.2L: 9579498 - 9581745 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0014179
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001273363.1 NP_001260292.1 1 - 2 549 58
NM_057760.5 NP_477108.1 1 - 2 549 58
GenBank mRNA BT029286.1 - 1 - 2 405 58

View Full Product Details