Assay Design



Gene Transcripts

Gene Symbol
dare
Entrez Gene ID
36203
Gene Name
defective in the avoidance of repellents
Gene Aliases
AR, CG12390, Dare, Dmel\CG12390, Dmel_CG12390, W8
Chromosome Location
Chr.2R: 11290994 - 11293174 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0015582
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_057802.6 NP_477150.1 1 - 2 364 82
GenBank mRNA AF168685.1 - 1 - 2 340 82
BT083432.1 - 1 - 2 361 82

View Full Product Details