Assay Design



Gene Transcripts

Gene Symbol
Cyt-b5-r
Entrez Gene ID
35008
Gene Name
Cytochrome b5-related
Gene Aliases
CG13279, Cyt-b, Cyt-b5, Cytb, Cytb5, Cytb5r, Dmel\CG13279, Dmel_CG13279, TU-36B, TU36B
Chromosome Location
Chr.2L: 16788774 - 16790721 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0000406
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001273585.1 NP_001260514.1 1 - 2 509 68
NM_057806.5 NP_477154.1 1 - 2 509 68
GenBank mRNA AY058287.1 - 1 - 2 497 68

View Full Product Details