Assay Design



Gene Transcripts

Gene Symbol
vkg
Entrez Gene ID
33726
Gene Name
viking
Gene Aliases
1209, 6072, CG16858, CT25584, ColIV, ColIValpha2, Coll IV, Coll IValpha2, DmColA2, Dmel\CG16858, Dmel_CG16858, Vkg, VkgC, alpha(IV)2/vkg, col4a2, coll-IV, coll. IV, collagen-IV, l(2)01209
Chromosome Location
Chr.2L: 5012144 - 5028191 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0016075
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001273142.1 NP_001260071.1 6 - 7 5016 61
NM_001273143.1 NP_001260072.1 6 - 7 5091 61
NM_057842.3 NP_477190.1 6 - 7 5037 61

View Full Product Details