Assay Design
Gene Transcripts
- Gene Symbol
- Dredd
- Entrez Gene ID
-
31011
- Gene Name
- Death related ced-3/Nedd2-like caspase
- Gene Aliases
-
CASP8_DROME, CG7486, DCP-2, DCP-2/DREDD, DCP2, DREDD, Dcp-2, Dcp-2/Dredd, Dcp2, Dedd, Dmel\CG7486, Dmel_CG7486, Dredd/DCP-2, EG:115C2.9, Redd, anon-1BCa, dDredd, dcp-2/dredd, dredd
- Chromosome Location
-
Chr.X: 633622 - 635545 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0020381
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_057903.4 |
NP_477251.3 |
2 - 3 |
880 |
90 |
View Full Product Details