Assay Design



Gene Transcripts

Gene Symbol
Alas
Entrez Gene ID
37815
Gene Name
Aminolevulinate synthase
Gene Aliases
ALAS, CG3017, Dmel\CG3017, Dmel_CG3017, alas
Chromosome Location
Chr.2R: 23964772 - 23966757 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0020764
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_057933.5 NP_477281.1 1 - 2 1358 66
GenBank mRNA BT044432.1 - 1 - 2 1323 66
Y14576.1 - - 1262 66

View Full Product Details