Assay Design
Gene Transcripts
- Gene Symbol
- Acp32CD
- Entrez Gene ID
-
34531
- Gene Name
- Accessory gland protein 32CD
- Gene Aliases
-
CG4605, Dmel\CG4605, Dmel_CG4605
- Chromosome Location
-
Chr.2L: 11015620 - 11016571 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0023415
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_057980.5 |
NP_477328.2 |
1 - 2 |
803 |
85 |
View Full Product Details