Assay Design
Gene Transcripts
- Gene Symbol
- beta'COP
- Entrez Gene ID
-
45757
- Gene Name
- Coat Protein (coatomer) beta'
- Gene Aliases
-
CG6699, COPIbeta', Dmel\CG6699, Dmel_CG6699, beta'-COP, beta'Cop, dbeta'COP
- Chromosome Location
-
Chr.2L: 13378902 - 13382382 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0025724
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_080097.4 |
NP_524836.2 |
2 - 3 |
1094 |
69 |
GenBank mRNA |
BT023906.1 |
- |
2 - 3 |
1085 |
69 |
View Full Product Details