Assay Design



Gene Transcripts

Gene Symbol
Eip63F-1
Entrez Gene ID
47878
Gene Name
Ecdysone-induced protein 63F 1
Gene Aliases
63F-1, CBP12, CG15855, Ca[2+] binding protein, Dmel\CG15855, Dmel_CG15855, E63-1, E63F, Eip63-1, Eip63F1, Eip63Fa, eip63F-1
Chromosome Location
Chr.3L: 3909973 - 3935617 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0004910
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202117.2 NP_001189046.1 3 - 4 217 73
NM_080163.4 NP_524902.2 3 - 4 476 73
GenBank mRNA BT088940.1 - 3 - 4 455 73

View Full Product Details