Assay Design
Gene Transcripts
- Gene Symbol
- Spn38F
- Entrez Gene ID
-
49806
- Gene Name
- Serpin 38F
- Gene Aliases
-
CG9334, Dmel\CG9334, Dmel_CG9334, Sp3, Spn3, sp3
- Chromosome Location
-
Chr.2L: 20836621 - 20837985 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0028986
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_080217.4 |
NP_524956.2 |
3 - 4 |
998 |
76 |
View Full Product Details