Assay Design



Gene Transcripts

Gene Symbol
wun2
Entrez Gene ID
53558
Gene Name
wunen-2
Gene Aliases
BEST:CK02248, CG8805, CK02248, Dmel\CG8805, Dmel_CG8805, DrPAP2[G], LPP-like, N14, Pap2G, Tunen, WUN-like, Wun2, wun-2
Chromosome Location
Chr.2R: 9414342 - 9417846 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0041087
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001299319.1 NP_001286248.1 2 - 3 1417 59
NM_080252.4 NP_524991.1 2 - 3 1174 59
GenBank mRNA AF331162.1 - 2 - 3 1152 59

View Full Product Details