Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001169168.2 | NP_001162639.2 | 5 - 6 | 1451 | 78 |
NM_001169169.2 | NP_001162640.1 | 5 - 6 | 1445 | 78 | |
NM_001169170.2 | NP_001162641.1 | 4 - 5 | 1638 | 78 | |
NM_080300.3 | NP_525039.1 | 5 - 6 | 1451 | 78 | |
GenBank mRNA | AF277393.1 | - | 4 - 5 | 1196 | 78 |
BT064640.1 | - | 5 - 6 | 1443 | 78 |