Assay Design
Gene Transcripts
- Gene Symbol
- Cyp4ae1
- Entrez Gene ID
-
31193
- Gene Name
- Cytochrome P450-4ae1
- Gene Aliases
-
4ae1, CG10755, CYP4E4, Cyp4e4, Cypae1, Dmel\CG10755, Dmel_CG10755, EG:152A3.6, I, P-450, P450, TcD20, anon-2Ea
- Chromosome Location
-
Chr.X: 2179007 - 2180998 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0015036
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_080305.4 |
NP_525044.1 |
3 - 4 |
1241 |
78 |
GenBank mRNA |
AY058450.1 |
- |
3 - 4 |
1241 |
78 |
View Full Product Details