Assay Design



Gene Transcripts

Gene Symbol
pcx
Entrez Gene ID
31204
Gene Name
pecanex
Gene Aliases
CG3443, DMPCX, Dmel\CG3443, Dmel_CG3443, EG:30B8.4, fs(1)12-1012, fs(1)12-1743, fs(1)12-2135, fs(1)12-3014, fs(1)12-3102, fs(1)12-4169, fs(1)14-1153, fs(1)14-567, fs(1)A387, fs(1)M10, fs(1)pcx, mel(1)R, mel(1)R1
Chromosome Location
Chr.X: 2225296 - 2238045 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0003048
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001272253.1 NP_001259182.1 4 - 5 4839 57
NM_001272254.2 NP_001259183.2 4 - 5 4851 57
NM_080306.4 NP_525045.3 4 - 5 4839 57

View Full Product Details