Assay Design



Gene Transcripts

Gene Symbol
bi
Entrez Gene ID
31379
Gene Name
bifid
Gene Aliases
BI, CG3578, Dm-OMB, Dm-omb, Dmel\CG3578, Dmel_CG3578, OMB, Omb, Qd, T3, TBX2, dm-omb, l(1)bi, l(1)omb, omb
Chromosome Location
Chr.X: 4412856 - 4485647 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0000179
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169191.1 NP_001162662.1 7 - 8 2481 73
NM_001169192.2 NP_001162663.1 7 - 8 2448 73
NM_001272309.2 NP_001259238.1 7 - 8 2481 73
NM_080331.3 NP_525070.2 7 - 8 2108 73

View Full Product Details