Assay Design



Gene Transcripts

Gene Symbol
Mgat1
Entrez Gene ID
37323
Gene Name
Mannosyl (alpha-1,3-)-glycoprotein beta-1,2-N-acetylglucosaminyltransferase
Gene Aliases
CG13431, Dmel\CG13431, Dmel_CG13431, GlcNAc-TI, GlcNAcT1, MGAT1, dMGAT1, mgat1
Chromosome Location
Chr.2R: 20559156 - 20562331 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0034521
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080378.4 NP_525117.2 2 - 3 1455 88
GenBank mRNA AB176552.1 - 2 - 3 894 88
BT032838.1 - 2 - 3 1411 88
BT124781.1 - 3 - 4 2286 88

View Full Product Details