Assay Design
Gene Transcripts
- Gene Symbol
- TotF
- Entrez Gene ID
-
117461
- Gene Name
- Turandot F
- Gene Aliases
-
CG13963, CG31691, Dmel\CG31691, Dmel_CG31691, Tot F, tot
- Chromosome Location
-
Chr.2L: 19911815 - 19912377 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0044811
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_080519.4 |
NP_536780.3 |
2 - 2 |
360 |
59 |
GenBank mRNA |
BT122095.2 |
- |
2 - 2 |
315 |
59 |
View Full Product Details