Assay Design



Gene Transcripts

Gene Symbol
RhoGAP16F
Entrez Gene ID
32790
Gene Name
Rho GTPase activating protein at 16F
Gene Aliases
CG7122, Dmel\CG7122, Dmel_CG7122
Chromosome Location
Chr.X: 18093145 - 18097413 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0030893
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001258801.2 NP_001245730.1 2 - 3 1533 69
NM_001298471.1 NP_001285400.1 2 - 3 1533 69
NM_001298472.1 NP_001285401.1 3 - 4 2268 69
NM_133039.5 NP_573267.3 2 - 3 1594 69
NM_176755.3 NP_788928.2 3 - 4 1793 69
GenBank mRNA BT133383.1 - 3 - 4 1777 69

View Full Product Details