Assay Design



Gene Transcripts

Gene Symbol
Rip11
Entrez Gene ID
32850
Gene Name
Rab11 interacting protein
Gene Aliases
CG6606, Dmel\CG6606, Dmel_CG6606, dRip11, l(1)G0003, l(1)G0070, l(1)G0092, l(1)G0205, l(1)G0297, l(1)G0434
Chromosome Location
Chr.X: 18635642 - 18647648 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0027335
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001298482.1 NP_001285411.1 6 - 7 1828 71
NM_001298483.1 NP_001285412.1 6 - 7 3046 71
NM_133085.4 NP_573313.2 6 - 7 3128 71

View Full Product Details