Assay Design



Gene Transcripts

Gene Symbol
RhoGAP18B
Entrez Gene ID
32898
Gene Name
Rho GTPase activating protein at 18B
Gene Aliases
5-131, CG17015, CG42274, CG7481, CG7502, CG7531, Dmel\CG42274, Dmel_CG42274, whir
Chromosome Location
Chr.X: 19136107 - 19167082 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0261461
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144748.2 NP_001138220.1 4 - 5 1783 67
NM_001144749.2 NP_001138221.1 4 - 5 881 67
NM_001298497.1 NP_001285426.1 4 - 5 979 67
NM_133123.2 NP_573351.1 3 - 4 1400 67
NM_133124.4 NP_573352.2 5 - 6 3408 67
GenBank mRNA AY061332.1 - 3 - 4 1382 67
DQ486141.1 - 4 - 5 863 67
DQ486142.1 - 4 - 5 1779 67

View Full Product Details