Assay Design
Gene Transcripts
- Gene Symbol
- b6
- Entrez Gene ID
-
31124
- Gene Name
- CG3100-PA
- Gene Aliases
-
B6, CG3100, Dmel\CG3100, Dmel_CG3100, EG:9D2.1
- Chromosome Location
-
Chr.X: 1686624 - 1690187 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0024897
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_058017.5 |
NP_477365.1 |
2 - 3 |
785 |
67 |
GenBank mRNA |
BT050466.1 |
- |
2 - 3 |
783 |
67 |
View Full Product Details