Assay Design



Gene Transcripts

Gene Symbol
Pten
Entrez Gene ID
43991
Gene Name
Phosphatase and tensin homolog
Gene Aliases
CG5671, D.PTEN, DPTEN, Dmel\CG5671, Dmel_CG5671, PTEN, PTEN3, dPTEN, dPten, dpten, pten
Chromosome Location
Chr.2L: 10256319 - 10261049 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0026379
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_134320.3 NP_599147.1 11 - 12 2163 75
NM_134321.3 NP_599148.1 9 - 10 1933 75
GenBank mRNA AF161259.1 - 9 - 10 1944 75

View Full Product Details